Skip to Content
MilliporeSigma
All Photos(1)

Documents

DWK294010203

One Pack Contains: 100 Self-Adhesive Duran Youtility Bottle Labels (36 X 70Mm)

Synonym(s):

294010203

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 294010203

Application

identification

Features and Benefits

Printable self-adhesive; Wide – 40 to + 150 °C thermal performance range

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service