Skip to Content
MilliporeSigma
All Photos(2)

Documents

DWK293033508

Tamper-Proof Screw Caps For Wide Neck Square Bottles

neck Joints: (GL 60 H)

Synonym(s):

293033508

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

material

blue polypropylene screw cap

Joint

Joints: neck (GL 60 H)

packaging

pkg of 10 ea

manufacturer/tradename

293033508

Looking for similar products? Visit Product Comparison Guide

Application

Sterile liquid transfer between containers, Liquid sampling into bottles from larger containers, Port assemblies for spinner flasks, and cell culture vessels,

Features and Benefits

for lime soda bottle GL 60

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service