Skip to Content
MilliporeSigma
All Photos(2)

Documents

DWK292432875

Bottle Tag, For Gl 45 Duran® Media Bottles Bottle Tag

black silicone bottle

Synonym(s):

292432875

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.35

material

black silicone bottle

packaging

pkg of 20 ea

manufacturer/tradename

DURAN® 292432875

Looking for similar products? Visit Product Comparison Guide

General description

The colourful GL 45 Bottle Tags offer an elegant solution to the marking of laboratory bottles. The Bottle Tags can be easily and securely attached around the neck of the YOUTILITY bottle, even with the screw cap in place. The Bottle Tags feature a graspable tab that makes them easy to remove with bare or gloved hands.

Application

easy customisation and unambiguous bottle identification.

Features and Benefits

for GL 45 bottles

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service