Key Documents
DWK217311704
DURAN® wide mouth reaction flask with beaded rim
round bottom glass flask, flask capacity (50 mL), single-neck
Synonym(s):
217311704
About This Item
UNSPSC Code:
41121804
NACRES:
NB.17
material
round bottom glass flask
packaging
case of 10 ea
manufacturer/tradename
DURAN® 217311704
flask capacity
(50 mL)
General description
Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.
Features and Benefits
• Complies with DIN ISO 24450• Manufactured from borosilicate glass 3.3• Flat base means flasks can be set down without a supporting ring• The wide neck enables easy filling and cleaning• Fired-on, highly durable white ceramic
Legal Information
DURAN is a registered trademark of DWK Life Sciences