Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWK213543307

DURAN® powder funnel with short wide stem

glass funnel(s), funnel capacity (25 mL)

Synonym(s):

213543307

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

material

glass funnel(s)

packaging

case of 10 ea

manufacturer/tradename

DURAN® 213543307

funnel capacity

(25 mL)

Looking for similar products? Visit Product Comparison Guide

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service