Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWK213525709

Duran® Funnel Ribbed

Synonym(s):

213525709

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43
Product DWK213525709 is not currently sold in your country. Contact Technical Service

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 213525709

funnel diam.

150 mm

Looking for similar products? Visit Product Comparison Guide

Application

Decanting and filtration of liquids, The spillage of substances is avoided, Filtering, Boro 3.3

Features and Benefits

Due to its shape, it is suitable for liquid-solid filtration using round filter paper; Fired-on,

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service