Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWK213112401

Duran® Crystallizing Dish

cylinder capacity 20 mL

Synonym(s):

213112401

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.43

description

With Spout, With Print

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 213112401

cylinder capacity

20 mL

Looking for similar products? Visit Product Comparison Guide

Application

heating and cooling baths, crystallization, disposal by evaporation and sand baths for microscale experiments

Features and Benefits

heating and cooling baths, crystallization, disposal by evaporation and sand baths for microscale experiments

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service