Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D91403

Sigma-Aldrich

Diethylcarbamoyl chloride

97%

Synonym(s):

Chloroformic acid diethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCOCl
CAS Number:
Molecular Weight:
135.59
Beilstein:
506687
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

117-123 °C/133 mmHg (lit.)

mp

−32 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)C(Cl)=O

InChI

1S/C5H10ClNO/c1-3-7(4-2)5(6)8/h3-4H2,1-2H3

InChI key

OFCCYDUUBNUJIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Frazier et al.
Journal of applied microbiology, 126(1), 79-86 (2018-08-31)
Disulfiram (Antabuse™) and its metabolites formed in vivo were evaluated as antibacterial agents against thirty species of Gram-positive and Gram-negative bacteria. The synergistic potential of disulfiram (DSF) and metabolite diethyldithiocarbamate (DDTC) with approved antibiotics were also compared by isobologram (checkerboard) analysis.
[Determination of low levels of diethylcarbamoyl chloride in air. First results after inhalation in rat (author's transl)].
J P Anger et al.
Toxicological European research. Recherche europeenne en toxicologie, 3(5), 223-228 (1981-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service