Skip to Content
MilliporeSigma
All Photos(1)

Documents

D90008

Sigma-Aldrich

Diethyl azodicarboxylate

Synonym(s):

DEAD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCON=NCOOC2H5
CAS Number:
Molecular Weight:
174.15
Beilstein:
908662
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

form

liquid

impurities

<3% dichloromethane

refractive index

n20/D 1.43 (lit.)

bp

106 °C/13 mmHg (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)\N=N\C(=O)OCC

InChI

1S/C6H10N2O4/c1-3-11-5(9)7-8-6(10)12-4-2/h3-4H2,1-2H3/b8-7+

InChI key

FAMRKDQNMBBFBR-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used in Mitsunobu and Diels-Alder reactions, as well as in allylic amination of alkenes.

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Expl. 1.1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

1 - Explosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Journal of Organic Chemistry, 58, 5261-5261 (1993)
The Journal of Organic Chemistry, 58, 4979-4979 (1993)
F K Brown et al.
Journal of medicinal chemistry, 37(5), 674-688 (1994-03-04)
Systematic modification of the presumed P1 side chain in a series of (carboxyalkyl)amino-based inhibitors of matrix metalloproteinases enabled identification of the 2-(1,3-dihydro-1,3-dioxo-2H-benz[f]isoindol-2-yl)ethyl group as a preferred substituent imparting potent inhibition of the enzymes collagenase and gelatinase. It was subsequently found
Jetze Visser et al.
Nature communications, 6, 6933-6933 (2015-04-29)
Despite intensive research, hydrogels currently available for tissue repair in the musculoskeletal system are unable to meet the mechanical, as well as the biological, requirements for successful outcomes. Here we reinforce soft hydrogels with highly organized, high-porosity microfibre networks that
Fany Reffuveille et al.
Antimicrobial agents and chemotherapy, 58(9), 5363-5371 (2014-07-02)
Biofilm-related infections account for at least 65% of all human infections, but there are no available antimicrobials that specifically target biofilms. Their elimination by available treatments is inefficient since biofilm cells are between 10- and 1,000-fold more resistant to conventional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service