Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D86353

Sigma-Aldrich

Diethyl 4-aminobenzylphosphonate

99%

Synonym(s):

4-(Diethylphosphonomethyl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
243.24
Beilstein:
2941613
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

99%

reaction suitability

reaction type: C-C Bond Formation

mp

90-94 °C (lit.)

SMILES string

CCOP(=O)(Cc1ccc(N)cc1)OCC

InChI

1S/C11H18NO3P/c1-3-14-16(13,15-4-2)9-10-5-7-11(12)8-6-10/h5-8H,3-4,9,12H2,1-2H3

InChI key

ZVAYUUUQOCPZCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guowei Jiang et al.
Bioorganic & medicinal chemistry letters, 21(17), 5098-5101 (2011-04-15)
Autotaxin (ATX) is an attractive target for the anticancer therapeutics that inhibits angiogenesis, invasion and migration. ATX is an extracellular lysophospholipase D that hydrolyzes lysophosphatidylcholine to form the bioactive lipid lysophosphatidic acid. The aromatic phosphonate S32826 was the first described

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service