Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D76404

Sigma-Aldrich

2,5-Dichloro-p-xylene

97%

Synonym(s):

1,4-Dichloro-2,5-dimethylbenzene, 2,5-Dichloro-1,4-dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H2Cl2
CAS Number:
Molecular Weight:
175.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

222 °C (lit.)

mp

69-70 °C (lit.)

SMILES string

Cc1cc(Cl)c(C)cc1Cl

InChI

1S/C8H8Cl2/c1-5-3-8(10)6(2)4-7(5)9/h3-4H,1-2H3

InChI key

UTGSRNVBAFCOEU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service