Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D74509

Sigma-Aldrich

2,6-Dichlorostyrene

99%

Synonym(s):

2,6-Dichlorovinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H3Cl2
CAS Number:
Molecular Weight:
173.04
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.574 (lit.)

bp

88-90 °C/8 mmHg (lit.)

density

1.267 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1cccc(Cl)c1C=C

InChI

1S/C8H6Cl2/c1-2-6-7(9)4-3-5-8(6)10/h2-5H,1H2

InChI key

YJCVRMIJBXTMNR-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio A Ceacero-Vega et al.
The journal of physical chemistry. A, 111(28), 6406-6419 (2007-06-27)
The molecular structure of 2,6-dichlorostyrene has been analyzed at MP2 and DFT levels using different basis sets concluding in a nonplanar geometry. The influence of either the level of theory or the nature of the substituent has been assessed. The
I Makhniashvili et al.
Fresenius' journal of analytical chemistry, 367(6), 568-571 (2001-02-28)
Conditions for a gas-chromatographic determination of vapors and aerosols of p-chlorostyrene and 2,6-dichlorostyrene contained in workplace air samples were determined. The method is based on the adsorption of p-chlorostyrene and 2,6-dichlorostyrene on activated charcoal and fiberglass, desorption with toluene and

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service