Assay
98%
refractive index
n20/D 1.442 (lit.)
bp
117-119 °C (lit.)
mp
−80 °C (lit.)
density
1.107 g/mL at 25 °C (lit.)
SMILES string
CC(Cl)C(C)Cl
InChI
1S/C4H8Cl2/c1-3(5)4(2)6/h3-4H,1-2H3
InChI key
RMISVOPUIFJTEO-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service