Skip to Content
MilliporeSigma
All Photos(1)

Documents

D5627

Sigma-Aldrich

p-Diazo-N,N-diethyl-m-toluidine hemi(zinc chloride) salt

practical grade

Synonym(s):

4-Diethylamino-2-methylbenzenediazonium chloride zinc chloride double salt, Diazo 575F

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H16N3Cl · 1/2ZnCl2
CAS Number:
Molecular Weight:
293.87
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

practical grade

SMILES string

Cl[Zn--](Cl)(Cl)Cl.CCN(CC)c1ccc([N+]#N)c(C)c1.CCN(CC)c2ccc([N+]#N)c(C)c2

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service