Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D41708

Sigma-Aldrich

2,6-Dibromo-4-methylphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H2(CH3)OH
CAS Number:
Molecular Weight:
265.93
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

49-50 °C (lit.)

SMILES string

Cc1cc(Br)c(O)c(Br)c1

InChI

1S/C7H6Br2O/c1-4-2-5(8)7(10)6(9)3-4/h2-3,10H,1H3

InChI key

FIGPGTJKHFAYRK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service