Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D25206

Sigma-Aldrich

4,4′-Diaminostilbene dihydrochloride

95%

Synonym(s):

4,4′-Vinylenedianiline dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH=CHC6H4NH2 · 2HCl
CAS Number:
Molecular Weight:
283.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

powder

mp

>300 °C (lit.)

SMILES string

Cl.Cl.Nc1ccc(cc1)\C=C\c2ccc(N)cc2

InChI

1S/C14H14N2.2ClH/c15-13-7-3-11(4-8-13)1-2-12-5-9-14(16)10-6-12;;/h1-10H,15-16H2;2*1H/b2-1+;;

InChI key

QHDAFTKIEDDTPV-SEPHDYHBSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service