Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D221406

Sigma-Aldrich

1-Dodecanethiol

98%

Synonym(s):

n-Dodecyl mercaptan, Dodecyl mercaptan, Lauryl mercaptan, Mercaptan C12, NDM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11SH
CAS Number:
Molecular Weight:
202.40
Beilstein:
969337
EC Number:
MDL number:

vapor density

7 (vs air)

Assay

98%

refractive index

n20/D 1.459 (lit.)

bp

266-283 °C (lit.)

density

0.845 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCS

InChI

1S/C12H26S/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

InChI key

WNAHIZMDSQCWRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service