All Photos(1)
About This Item
Linear Formula:
(O2N)2C6H3CN
CAS Number:
Molecular Weight:
193.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
form
powder
mp
126-130 °C (lit.)
SMILES string
[O-][N+](=O)c1cc(cc(c1)[N+]([O-])=O)C#N
InChI
1S/C7H3N3O4/c8-4-5-1-6(9(11)12)3-7(2-5)10(13)14/h1-3H
InChI key
SSDNULNTQAUNFQ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jia Wei Peng et al.
Molecules (Basel, Switzerland), 25(20) (2020-10-24)
In the present study, we aimed to obtain a high yield and productivity for glucosamine using a low-cost solid-state culture with Aspergillus sydowii BCRC 31742. The fermentation conditions, such as inoculum biomass, moisture content, and supplemental volume and mineral salt
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service