Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D183806

Sigma-Aldrich

2,5-Dimethylpyrrolidine, mixture of cis and trans

93%, technical grade

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$373.00

$373.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$373.00

About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$373.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical grade

Assay

93%

form

liquid

refractive index

n20/D 1.4299 (lit.)

bp

105-106 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

SMILES string

CC1CCC(C)N1

InChI

1S/C6H13N/c1-5-3-4-6(2)7-5/h5-7H,3-4H2,1-2H3

InChI key

ZEBFPAXSQXIPNF-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service