Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D158801

Sigma-Aldrich

DBE-5 dibasic ester

99%

Synonym(s):

Dimethyl glutarate, Glutaric acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCO(CH2)3COOCH3
CAS Number:
Molecular Weight:
160.17
Beilstein:
1771444
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.424 (lit.)

bp

93-95 °C/13 mmHg
96-103 °C/15 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCC(=O)OC

InChI

1S/C7H12O4/c1-10-6(8)4-3-5-7(9)11-2/h3-5H2,1-2H3

InChI key

XTDYIOOONNVFMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DuPont product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Misa Kuroki et al.
iScience, 23(1), 100786-100786 (2020-01-07)
Metabolic switching and rewiring play a dynamic role in programmed cell differentiation. Many pathogenic microbes need to survive in nutrient-deficient conditions and use the glyoxylate cycle, an anaplerotic pathway of the tricarboxylic acid cycle, to produce carbohydrates. The plant pathogenic
L Alvarez et al.
Drug and chemical toxicology, 18(4), 295-314 (1995-11-01)
Dibasic esters (DBE) are a mixture of 3 carboxylic acid esters which are used in the paint and coatings industry. In this study, groups of pregnant Crl:CD BR rats were exposed to either 0.16, 0.4, or 1.0 mg DBE/L by
J P Turner et al.
British journal of pharmacology, 104(2), 445-451 (1991-10-01)
1. The effects of L-glutamate diethyl ester (GDEE) HCl, glutarate diethyl ester (GlrDEE) and glutarate dimethyl ester (GlrDME) on depolarizing responses to alpha-amino-3-hydroxy-5- methyl-4-isoxazolepropionate (AMPA), kainate (Kain), N-methyl-D-aspartate (NMDA) and quisqualate (Quis), and spontaneous paroxysmal discharges (SPDs) were examined. Experiments
M S Bogdanffy et al.
Drug metabolism and disposition: the biological fate of chemicals, 19(1), 124-129 (1991-01-01)
Inhalation exposure of rats to dibasic esters revealed lesions of the nasal olfactory epithelium similar to those observed with other ester solvents. Female rats are more sensitive to these effects than are male rats. It has been proposed that carboxylesterase
J B Morris et al.
Toxicology and applied pharmacology, 108(3), 538-546 (1991-05-01)
Inhalation exposure of the male and female rat to high concentrations of a mixture of the dibasic esters dimethyl succinate (DMS), dimethyl glutarate (DMG), and dimethyl adipate (DMA) results in mild olfactory toxicity. This response is thought to be due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service