Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D104957

Sigma-Aldrich

Dibenzosuberane

98%

Synonym(s):

10,11-Dihydro-5H-dibenzo[a,d]cycloheptene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14
CAS Number:
Molecular Weight:
194.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

bp

160-165 °C/2 mmHg (lit.)

mp

74-76 °C (lit.)

SMILES string

C1Cc2ccccc2Cc3ccccc13

InChI

1S/C15H14/c1-3-7-14-11-15-8-4-2-6-13(15)10-9-12(14)5-1/h1-8H,9-11H2

InChI key

PJQCANLCUDUPRF-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A T Evans et al.
Biochemical pharmacology, 48(3), 613-616 (1994-08-03)
There is continuing interest in the antimicrobial effects of tricyclic neuroleptics and antidepressants, particularly against trypanosomatid protozoa, but few studies have attempted to dissociate antiparasitic actions from effects on the central nervous system. In this report, we have measured the
H Szumiło et al.
Acta poloniae pharmaceutica, 47(3-4), 11-15 (1990-01-01)
An influence of several adsorbents and the composition of the eluent mixtures on the separation of dibenzoazepine and dibenzocycloheptadiene derivatives was examined by thin-layer chromatography. The best system (RP-18 and methanol-buffer solution pH 8.5 19:1) was employed for the separation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service