Skip to Content
MilliporeSigma
All Photos(1)

Documents

CVT00136

Sigma-Aldrich

Methyl 5-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylate

Synonym(s):

5-Methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9F3N2O2
Molecular Weight:
222.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

CC1=C(C(OC)=O)C=NN1CC(F)(F)F

InChI

1S/C8H9F3N2O2/c1-5-6(7(14)15-2)3-12-13(5)4-8(9,10)11/h3H,4H2,1-2H3

InChI key

AAWMDUPUYVKILO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service