Key Documents
CLS69474LBO
Pyrex® resin reaction kettle bottom
capacity 4,000 mL
Synonym(s):
pyrex kettle, reaction equipment, reaction flasks
Select a Size
4 EA
$2,520.00
$2,520.00
Please contact Customer Service for Availability
Select a Size
Change View
4 EA
$2,520.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$2,520.00
Please contact Customer Service for Availability
material
borosilicate glass
wide-mouth
sterility
non-sterile
feature
closure type lid
packaging
case of 4
manufacturer/tradename
Corning 6947-4LBO
availability
not available in EU
capacity
4,000 mL
General description
Flange is finely ground for a tight seal.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)