Key Documents
CLS53202L
Pyrex® heavy-wall graduated filtering flask
capacity 2,000 mL
Synonym(s):
conical flasks, filter flasks, filtering flask, filtering flasks, flask filter, Corning® 5320
Select a Size
12 EA
$2,310.00
$2,310.00
Please contact Customer Service for Availability
Select a Size
Change View
12 EA
$2,310.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$2,310.00
Please contact Customer Service for Availability
material
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper9
description
without hose connection
sterility
non-sterile
feature
closure type Rubber stopper
packaging
pack of 1
case of 12
manufacturer/tradename
Corning 5320-2L
volume measuring range
600-2,000 mL
capacity
2,000 mL
General description
Blown in special molds to give maximum strength. Graduated to show approximate capacity.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)