Documents
CLS5100500
Pyrex® wide-mouth Erlenmeyer flask
capacity 500 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5100
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
wide-mouth
Joint
joint stopper10
sterility
non-sterile
feature
cap: no
subdivision 50
packaging
case of 36
pack of 6
manufacturer/tradename
Corning 5100-500
volume measuring range
100-500 mL
capacity
500 mL
General description
  • Pyrex® wide-mouth Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
  • Flasks have durable white enamel graduations to show approximate capacity and an extra-large marking spot.
  • Features a heavy-duty rim.
  • Ideal for use as a titration flask.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.