CLS396570
Pyrex® aseptic filling bell
hose outlet O.D. 11.0 mm, O.D. × H 73 mm × 130 mm
Synonym(s):
Corning® 3965
Select a Size
$224.00
Select a Size
About This Item
$224.00
Recommended Products
description
for use with bottles
manufacturer/tradename
Corning 3965-70
O.D. × H
73 mm × 130 mm
hose outlet O.D.
11.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service