Skip to Content
MilliporeSigma
All Photos(1)

Documents

CF0029

Sigma-Aldrich

4-Ethoxycarbonylphenoxytetrafluoropropionic acid Cs salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H9F4O5Cs
MDL number:
UNSPSC Code:
12352101

form

solid

reaction suitability

reaction type: C-C Bond Formation

Application

4-Ethoxycarbonylphenoxytetrafluoropropionic acid Cs salt is as a less hygroscopic, crystalline source of 4-ethoxycarbonylphenoxytetrafluoropropionic acid. The ester group can be used to further build up molecular complexity.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service