Skip to Content
MilliporeSigma
All Photos(1)

Documents

CF0023

Sigma-Aldrich

4-Bromophenoxytetrafluoethyl trimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H13BrF4OSi
CAS Number:
MDL number:
UNSPSC Code:
12352101

form

liquid

reaction suitability

reaction type: C-C Bond Formation

Application

After being activated with silicophilic activators, like fluorides, alkoxides or carboxylates, 4-bromophenoxytetrafluoroethyl trimethylsilane serves as a nucleophilic source of the 4-bromophenoxytetrafluoroethyl moiety in reaction with electrophiles such as aromatic aldehydes. The aromatic bromide can be used to build up molecular complexity using Pd-catalyzed cross couplings.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service