Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CF0007

Sigma-Aldrich

Alcohol Togni-(4-EtCO2-PhOCF2CF2)-reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H19F4IO4
CAS Number:
MDL number:
UNSPSC Code:
12352200

form

solid

reaction suitability

reaction type: C-C Bond Formation

Application

Alcohol Togni-(4-EtCO2-PhOCF2CF2)-reagent is an electrophilic hypervalent iodine-fluoroalkyl reagent introducing the 4-ethoxycarbonyl phenoxytetrafluoroethyl fragment. The ester group can be transformed to other derivatives, building additional molecular complexity.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service