Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS015019

Sigma-Aldrich

2-methylpropanimidamide hydrochloride

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11ClN2
Molecular Weight:
122.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

Cl.CC(C)C(N)=N

InChI

1S/C4H10N2.ClH/c1-3(2)4(5)6;/h3H,1-2H3,(H3,5,6);1H

InChI key

VWXLCWNPSOUPPE-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rashmi S Tupe et al.
Molecular biology reports, 47(1), 521-531 (2019-11-14)
Diabetic complications are associated with the glycation and formation of advanced glycation end products (AGEs) which leads to structural modifications of biomolecules further affecting cells. Carbonyl compounds such as methylglyoxal and glyceraldehyde-3-phosphate are highly reactive and form an elevated amount
Wei Wang et al.
The Journal of pharmacy and pharmacology, 72(11), 1645-1656 (2020-08-04)
Chamomile has long been used as a medicinal plant due to its antioxidative and anti-inflammatory activity. Apigenin-7-O-glucoside (AG) is one of the major ethanol extract components from chamomile; however, the underlying mechanism remains unclear. In this study, the antioxidant potential
Yun-Feng Zheng et al.
Journal of agricultural and food chemistry, 68(49), 14684-14697 (2020-11-26)
Perillae Folium (PF), which is extensively used as a dietary vegetable and medicinal herb, contains two varietal forms corresponding to purple perilla leaf (Perilla frutescens var. crispa) and green perilla leaf (Perilla frutescens var. frutescens). However, the components and efficacy
Lovemore Nkhata Malunga et al.
Food chemistry, 167, 311-319 (2014-08-26)
The effect of xylanase type (Trichoderma viride or Neocallimastix patriciarum) and graded ethanol fractionation on the antioxidant capacity (AOC) of arabinoxylan oligosaccharides (AXOS) obtained from wheat aleurone was investigated. AXOS yields were higher using N. patriciarum (62%) than T. viride
Mira Rosenblat et al.
BioFactors (Oxford, England), 41(4), 273-288 (2015-08-01)
Atherogenesis is associated with macrophage cholesterol and oxidized lipids accumulation and foam cell formation. However, two other major lipid-metabolizing cell classes, namely intestinal and liver cells, are also associated with atherogenesis. This study demonstrates that manipulations of cellular oxidative stress

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service