Skip to Content
MilliporeSigma
All Photos(1)

Documents

CDS010752

Sigma-Aldrich

1H,1H,2H,2H-Perfluorodecyltrimethoxysilane

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13F17O3Si
CAS Number:
Molecular Weight:
568.30
MDL number:
UNSPSC Code:
31201707
PubChem Substance ID:

form

liquid

reaction suitability

core: silicon
reagent type: catalyst

SMILES string

CO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OC)OC

InChI

1S/C13H13F17O3Si/c1-31-34(32-2,33-3)5-4-6(14,15)7(16,17)8(18,19)9(20,21)10(22,23)11(24,25)12(26,27)13(28,29)30/h4-5H2,1-3H3

InChI key

HJIMAFKWSKZMBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

390.2 °F

Flash Point(C)

199 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeonghoon Han et al.
Physical review. E, 101(4-1), 043108-043108 (2020-05-20)
When a water drop impinges on a flat superhydrophobic surface, it bounces off the surface after a certain dwelling time, which is determined by the Rayleigh inertial-capillary timescale. Recent works have demonstrated that this dwelling time (i.e., contact time) is
Lanlan Hou et al.
ACS nano, 13(4), 4124-4132 (2019-03-19)
Anisotropic interfaces with opposite properties provide numerous unusual physical chemical properties that have played irreplaceable roles in broad domains. Here, we rationally designed an anisotropic Janus membrane with opposite wettability and special interpenetrating interface microstructure, which shows a unidirectional liquid
Xiao Yan et al.
ACS nano, 13(7), 8169-8184 (2019-07-03)
With the recent advances in surface fabrication technologies, condensation heat transfer has seen a renaissance. Hydrophobic and superhydrophobic surfaces have all been employed as a means to enhance condensate shedding, enabling micrometric droplet departure length scales. One of the main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service