Skip to Content
MilliporeSigma
All Photos(1)

Documents

CDS000253

Sigma-Aldrich

4,4′-Diamino[1,1′-biphenyl]-3,3′-diol

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O2
CAS Number:
Molecular Weight:
216.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

Nc1ccc(cc1O)-c2ccc(N)c(O)c2

InChI

1S/C12H12N2O2/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6,15-16H,13-14H2

InChI key

ZGDMDBHLKNQPSD-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Nagao et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 33(1), 35-39 (2017-01-11)
Two sulfonated polyimide (SPI) thin films were prepared with water and THF/water mixed solvents, respectively. The SPI thin film prepared with THF/water showed more than 5 times higher proton conductivity than that prepared with water mixed solvent at low relative
Beat J Brüschweiler et al.
Regulatory toxicology and pharmacology : RTP, 88, 214-226 (2017-06-29)
Azo dyes represent the by far most important class of textile dyes. Their biotransformation by various skin bacteria may release aromatic amines (AAs) which might be dermally absorbed to a major extent. Certain AAs are well known to have genotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service