Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CBR00329

Sigma-Aldrich

4-Hydrazinoquinazoline

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N4
CAS Number:
Molecular Weight:
160.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

NNC1=NC=NC2=CC=CC=C12

InChI

1S/C8H8N4/c9-12-8-6-3-1-2-4-7(6)10-5-11-8/h1-5H,9H2,(H,10,11,12)

InChI key

QVKNQXCOGYNFPC-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hemato-encephalic barrier function in strokes in patients with arterial hypertension under drug correction].
V V Dunaev et al.
Likars'ka sprava, (5-6)(5-6), 124-126 (1995-05-01)
Kazuhiro Yamazaki et al.
Journal of enzyme inhibition and medicinal chemistry, 28(1), 143-147 (2011-12-16)
Poly(ADP-ribose) polymerase (PARP)-1 inhibitor has been suggested to attenuate the ischemia-reperfusion injury. We investigated the protective effect of the cardioplegia with a PARP-1 inhibitor, 4-hydoxyquinazoline (4-HQ), against myocardial ischemia-reperfusion injury. Isolated rat hearts were perfused on a Langendorff apparatus and
Maher A El-Hashash et al.
Global journal of health science, 4(1), 174-183 (2012-09-18)
The reactions of 2-ethoxy-4-hydrazinoquinazoline 2 with diethyl oxalate and ethyl chloroacetate gave 6-ethoxy-2H-[1,2,4]triazino[4,3-c]quinazoline-3,4-dione 3 and 6-ethoxy-2,3-dihydro-4H-[1,2,4]triazino[4,3-c]quinazolin-4-one 4 respectively. A series of 5-ethoxy-2-X-[1,2,4]triazolo[1, 5-c]quinazolines 5a-d was also produced by reacting 2 with the acid chlorides namely: benzoyl, crotonyl, cinnamyl and 2-furoyl
Andriy G Golub et al.
European journal of medicinal chemistry, 58, 258-264 (2012-11-07)
Hepatitis C virus (HCV) NS5B polymerase is a key target for the development of anti-HCV drugs. Here we report on the identification of novel allosteric inhibitors of HCV NS5B through a combination of structure-based virtual screening and in vitro NS5B
V V Dunaev et al.
Ukrainskii biokhimicheskii zhurnal (1978), 65(3), 118-120 (1993-05-01)
It is established that KS-79 compound possesses the high level of antioxidant activity and realizes its antioxidant effect at initial stages of free-radical hydrogen peroxide oxidation of lipids. Due to this ability it differs from the oxidants of direct-type action.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service