Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C78607

Sigma-Aldrich

Cholesteryl palmitate

97%

Synonym(s):

5-Cholestene 3-palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H76O2
CAS Number:
Molecular Weight:
625.06
Beilstein:
2342867
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

form

powder

optical activity

[α]27/D −24°, c = 2 in chloroform

mp

74-77 °C (lit.)

transition temp

cholesteric phase to smectic phase 78.5 °C
crystalline phase to cholesteric phase 79 °C
cholesteric phase to isotropic phase 83 °C

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCC(=O)O[C@H]1CC[C@]2(C)C3CC[C@]4(C)C(CCC4C3CC=C2C1)[C@H](C)CCCC(C)C

InChI

1S/C43H76O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-23-41(44)45-36-28-30-42(5)35(32-36)24-25-37-39-27-26-38(34(4)22-20-21-33(2)3)43(39,6)31-29-40(37)42/h24,33-34,36-40H,7-23,25-32H2,1-6H3/t34-,36+,37+,38-,39+,40+,42+,43-/m1/s1

InChI key

BBJQPKLGPMQWBU-JADYGXMDSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Ludmir et al.
Obstetrics and gynecology, 72(3 Pt 1), 360-362 (1988-09-01)
Amniotic fluid cholesteryl palmitate, as measured by thin-layer chromatography, appears to be a very sensitive and specific predictor for the risk of respiratory distress syndrome (RDS) in newborns of normal pregnancies. To assess the effect of diabetes mellitus on fetal
A L MacKay et al.
Biochimica et biophysica acta, 601(1), 22-33 (1980-09-02)
Dispersions (50 wt% in water) of sphingomyelin/cholesteryl palmitate (95 : 5 mol%) have been studied by 2H- and 31P-NMR spectroscopy between 25 and 60 degrees C. The deuterated esters, cholesteryl palmitate-d31 and cholesteryl palmitate-16, 16, 16-d3, were used for 2H-NMR
J Ludmir et al.
American journal of obstetrics and gynecology, 157(1), 84-88 (1987-07-01)
Cholesteryl palmitate was measured by thin-layer chromatography in 98 amniotic fluid samples obtained from pregnancies ranging from 15 to 40 weeks. Pregnancies complicated by diabetes mellitus or Rh sensitization were excluded. Cholesteryl palmitate concentration increased with gestational age from values
M Rosseneu et al.
Atherosclerosis, 108(2), 127-136 (1994-08-01)
The European Atherosclerosis Research Study (EARS) is a study of the biological expression of a paternal history of premature coronary heart disease. The study was carried out in 14 centres in 11 European countries, where the offspring of fathers who
S M Innis et al.
The Journal of nutrition, 125(1), 73-81 (1995-01-01)
Milk fatty acids contain 20-30% palmitic acid (16:0), with approximately 70% of the 16:0 esterified to the sn-2 position of the milk triacylglycerol. Formulae containing vegetable and oleo oils contain different amounts of 16:0, but all have unsaturated fatty acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service