Skip to Content
MilliporeSigma
All Photos(1)

Documents

C44702

Sigma-Aldrich

5-Chloro-2-hydroxybenzophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
232.66
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

96-98 °C (lit.)

SMILES string

Oc1ccc(Cl)cc1C(=O)c2ccccc2

InChI

1S/C13H9ClO2/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8,15H

InChI key

OMWSZDODENFLSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanne Frederiksen et al.
International journal of hygiene and environmental health, 220(2 Pt A), 244-253 (2016-09-18)
Experimental studies indicate that some chemicals with UV blocking properties (known as UV filters) can act as endocrine disruptors. UV filters are used in sunscreens and other cosmetic- and personal care products, as well as in other consumer products such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service