Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C34308

Sigma-Aldrich

5-Chlorodibenzosuberane

96%

Synonym(s):

5-Chloro-10,11-dihydro-5H-dibenzo[a,d]cycloheptene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13Cl
CAS Number:
Molecular Weight:
228.72
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

106-107 °C (lit.)

SMILES string

ClC1c2ccccc2CCc3ccccc13

InChI

1S/C15H13Cl/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8,15H,9-10H2

InChI key

QPERNSDCEUTOTE-UHFFFAOYSA-N

Application

Reagent for introduction of the 5-dibenzosuberyl moiety as a modulator for anticancer drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Dhainaut et al.
Journal of medicinal chemistry, 35(13), 2481-2496 (1992-06-26)
A series of 70 triazine derivatives have been synthesized and tested for their capacity to modulate multidrug resistance (MDR) in DC-3F/AD and KB-A1 tumor cells in vitro, in comparison with verapamil (VRP), a calcium channel antagonist currently used in therapy
Miki Sato et al.
The Analyst, 142(23), 4560-4569 (2017-11-09)
The detection limits and photoionization thresholds of polycyclic aromatic hydrocarbons and their chlorides and nitrides on the water surface are examined using laser two-photon ionization and single-photon ionization, respectively. The laser two-photon ionization methods are highly surface-selective, with a high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service