Skip to Content
MilliporeSigma
All Photos(1)

Documents

C25406

Sigma-Aldrich

2-Chlorobenzotrichloride

96%

Synonym(s):

α,α,α,2-Tetrachlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CCl3
CAS Number:
Molecular Weight:
229.92
Beilstein:
2046639
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

refractive index

n20/D 1.583 (lit.)

bp

260-264 °C (lit.)

mp

29-31 °C (lit.)

density

1.508 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1C(Cl)(Cl)Cl

InChI

1S/C7H4Cl4/c8-6-4-2-1-3-5(6)7(9,10)11/h1-4H

InChI key

MFHPYLFZSCSNST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service