Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

C24191

Sigma-Aldrich

4-Chlorobenzenesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SO2NH2
CAS Number:
Molecular Weight:
191.64
Beilstein:
1308891
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product C24191, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

98%

form

powder

mp

140-144 °C (lit.)

SMILES string

NS(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C6H6ClNO2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,(H2,8,9,10)

InChI key

HHHDJHHNEURCNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C R Clark et al.
Epilepsia, 31(4), 474-479 (1990-07-01)
The anticonvulsant and toxic properties of 4-chlorobenzenesulfonamide (ADD 55051) were compared with phenytoin (PHT), phenobarbital (PB), ethosuximide (ESM), and valproate (VPA). These compounds were evaluated in mice and rats using well-standardized anti-convulsant test procedures. The results indicate that ADD 55051
Determination of the content of 2-chlorobenzenesulphonamide and bis(p-chlorophenyl) sulphone in 4-chlorobenzenesulphonamide.
E R Ventre
Journal of chromatography, 333(1), 253-255 (1985-09-27)
Yohei Munei et al.
Bioorganic & medicinal chemistry letters, 21(1), 141-144 (2010-12-07)
We proposed a novel QSAR (quantitative structure-activity relationship) procedure called LERE (linear expression by representative energy terms)-QSAR involving molecular calculations such as ab initio fragment molecular orbital and generalized Born/surface area ones. We applied LERE-QSAR to two datasets for the
Padmakar V Khadikar et al.
Bioorganic & medicinal chemistry letters, 15(4), 931-936 (2005-02-03)
A novel use of NMR chemical shift of the SO(2)NH(2) protons (in dioxane as solvent) as a molecular descriptor is described for modeling the inhibition constant for benzene sulfonamides against the zinc enzyme carbonic anhydrase (CA, EC 4.2.1.1). The methodology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service