Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C110604

Sigma-Aldrich

Cyclooctylamine

97%

Synonym(s):

Aminocyclooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H15NH2
CAS Number:
Molecular Weight:
127.23
Beilstein:
2689120
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.482 (lit.)

bp

190 °C (lit.)

mp

−48 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

NC1CCCCCCC1

InChI

1S/C8H17N/c9-8-6-4-2-1-3-5-7-8/h8H,1-7,9H2

InChI key

HSOHBWMXECKEKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stelios Eleftheratos et al.
Bioorganic & medicinal chemistry letters, 20(14), 4182-4187 (2010-06-24)
Interaction of aminoadamantanes with the influenza A virus M2 proton channel was analyzed by docking simulations of a series of synthetic aminoadamantane derivatives, of differing binding affinity, into the crystal structure of the transmembrane (M2TM) pore. The pore blocking model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service