Skip to Content
MilliporeSigma
All Photos(1)

Documents

C108057

Sigma-Aldrich

N-Cyclohexyl-1,3-propanediamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NH(CH2)3NH2
CAS Number:
Molecular Weight:
156.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.482 (lit.)

bp

120-123 °C/20 mmHg (lit.)

mp

−17-−15 °C (lit.)

density

0.917 g/mL at 25 °C (lit.)

SMILES string

NCCCNC1CCCCC1

InChI

1S/C9H20N2/c10-7-4-8-11-9-5-2-1-3-6-9/h9,11H,1-8,10H2

InChI key

ITZPOSYADVYECJ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Huber et al.
Cancer research, 55(4), 934-943 (1995-02-15)
Spermine is often the most abundant polyamine in human tumors such as breast carcinomas. However, its specific role in tumor biology is still uncertain, since inhibitors of ornithine decarboxylase such as alpha-difluoromethylornithine depress cell growth while leaving spermine content mostly
Weiping Liu et al.
Chemical & pharmaceutical bulletin, 56(5), 659-662 (2008-05-03)
New JM118 (active form of satraplatin) analogues with N-cyclohexyl-1,3-propanediamine (N-chpda) as the carrier, cis-[Pt(N-chpda)X2] (X2=2Cl(-) (1), oxalate (2), malonate (3), 1,1-cyclobutanedicarboxylate (CBDCA) (3), and 3-hydroxy-1,1-cyclobutanedicarboxylate(HO-CBDCA) (4)), have been synthesized and characterized by elemental analysis and spectroscopic data along with X-ray
P J Chu et al.
European journal of pharmacology, 256(2), 155-160 (1994-04-21)
The facilitating or antagonizing effects of polyamine analogues on N-methyl-DL-aspartate (NMDLA)-induced seizures were investigated using mice. Intracerebroventricular injection of spermidine and spermine, but not putrescine, shortened the latency to appearance of clonic convulsion induced by subcutaneous administration of NMDLA. Injection
A Shirahata et al.
Biochemical pharmacology, 45(9), 1897-1903 (1993-05-05)
Several inhibitors of aminopropyltransferases, developed recently in this laboratory, were tested for their specificity by measuring their effects on six enzyme activities related to polyamine biosynthesis and interconversion. Two of them, trans-4-methylcyclohexylamine (4MCHA) and N-(3-aminopropyl)cyclohexylamine (APCHA), selectively and potently inhibited
Kazuhiro Nishimura et al.
The Biochemical journal, 385(Pt 3), 779-785 (2004-09-21)
To examine the roles of active hypusinated eIF5A (eukaryotic translation initiation factor 5A) and polyamines in cell proliferation, mouse mammary carcinoma FM3A cells were treated with an inhibitor of deoxyhypusine synthase, GC7 (N1-guanyl-1, 7-diaminoheptane), or with an inhibitor of ornithine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service