Key Documents
BR81919
BRAND® burette straight stopcock
for burette capacity 50 mL, glass key
Synonym(s):
Accessories, Burette stopcock, Burettes, Stopcocks
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
material
amber
glass key
packaging
pack of 1 ea
manufacturer/tradename
BRAND 81919
General description
BRAND burettes are always calibrated to deliver (TD, Ex).
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)