BR782091
BRAND® microplate BRANDplates®, cellGrade prem
size 96 wells, size 330 μL, flat bottom, black, with transparent bottom
Synonym(s):
BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade premium
About This Item
flat bottom
polystyrene
96 wells
skirt Skirt (F-bottom)
material
black wells
flat bottom
polystyrene
sterility
sterile
feature
lid
skirt Skirt (F-bottom)
packaging
pack of 50 ea
pkg of (individually packed)
manufacturer/tradename
BRAND 782091
size
330 μL
96 wells
working volume
330 μL
color
black, with transparent bottom
binding type
cell culture treated
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Optimal surfaces for optimal results
- Reliable quality from the cleanroom
- Tested by independent laboratories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service