BR781615
BRAND® microplate BRANDplates®, pureGrade
size 96 wells, size 350 μL, foil bottom, black
Synonym(s):
96-well, BRANDplates®, Microplates, medium binding, plates, pureGrade
Select a Size
$1,500.00
Select a Size
About This Item
foil bottom
polystyrene
96 wells
$1,500.00
Recommended Products
material
black wells
foil bottom
polystyrene
sterility
non-sterile
feature
lid: no
skirt Skirt (F-bottom)
UV transparent plates
packaging
pack of 50 ea
manufacturer/tradename
BRAND 781615
size
350 μL
96 wells
working volume
350 μL
color
black
suitability
suitable for (bacteriological assays)
binding type
medium binding surface
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Optimal surfaces for optimal results
- Tested by independent laboratories
- BRANDplates® pureGrade microtitration plates are manufactured from high purity, crystal-clear polystyrene.
- Cleanroom production ensures the best possible cleanliness.
- Medium binding, non-treated, non-sterile surface
- BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
- Best suited for homogenous assays, screening, and storage.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service