Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR705433

BRAND® Transferpette® multichannel, electronic pipette

AC/DC input 230 V AC, Australian plug, 8-channel, volume 5-100 μL

Synonym(s):

Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Transferpette® electronic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

8-channel
accuracy± 0.8%
accuracy± 0.8 μL
electronic
± 0.25% Coefficient of variation
± 0.25 μL Coefficient of variation

packaging

pack of 1 ea

manufacturer/tradename

BRAND 705433

volume

5-100 μL

battery power

rechargeable

Looking for similar products? Visit Product Comparison Guide

General description

Transferpette electronic micropipettes combine the globally proven properties of mechanical piston operated air displacement micropipettes by BRAND with the advantages of an electronic device. The Transferpette electronic is the first micropipette to be awarded the "Ergonomics Approved" certificate from the TÜV Rheinland. The special FKM V-rings allow effortless mounting and easy ejection of virtually all commercially available pipette tips.
  • Motor-driven to reduce the risk of RSI and imprecision/inaccuracy caused by operator variability
  • Five modes: Pipetting, Reverse pipetting, Mixing, Dispensing, Gel-electrophoresis
  • Easy and rapid calibration without tools
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).

For pipettes up to 300 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, device stand, TipBox, refill pack, reagent reservoir, installation tool, silicone grease, and 1 set of sealing rings

For pipettes up to 1250 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, support frame, shelf mount, 2x TipBox, reagent reservoir, installation tool, grease and 1 set of sealing rings.

Features and Benefits

• Ergonomic, handy design• Clear and intuitive operation• Up to 4,000 pipetting cycles on a single charge

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service