Key Documents
BR705220
BRAND® HandyStep® Touch and HandyStep® Touch S Charging Stand
Select a Size
1 EA
$581.00
$581.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$581.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
$581.00
Please contact Customer Service for Availability
manufacturer/tradename
BRAND 705220
General description
The charging stand for the HandyStep touch saves space on the laboratory bench and securely holds the device even with a PD-Tip II attached. It features inductive charging technology, which enables charging without cable clutter. Simply place the HandyStep touch into the stand, and the charging process begins.
The blue LED status indicator at the base of the charging stand shows the charging status:
LED lit: Device is charging
LED not lit: Device is fully charged
The charging stand can be used for both the HandyStep touch and the HandyStep touch S.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
HandyStep is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)