Key Documents
BR62010
BRAND® PFA sample jar with screw cap
capacity 60 mL
Synonym(s):
Jars, PFA
About This Item
UNSPSC Code:
41121800
Pricing and availability is not currently available.
material
PFA
manufacturer/tradename
BRAND 62010
capacity
60 mL
fitting
GL 40
General description
For sample collection, transport and storage. PFA screw cap with molded sealing ring for a positive seal. Autoclavable, easy to clean, withstand temperatures from -200 °C to 260 °C.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)