Documents
BR462723
BRAND® rubber conical gasket for filter funnels and filter flasks
size 35 mm
Synonym(s):
Filter, Rubber gasket
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
EPDM rubber
packaging
pack of 10 ea
manufacturer/tradename
BRAND 462723
size
35 mm
Related Categories
General description
Conical gaskets to place between filter funnel and filter flask.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG