Key Documents
BR45620
BRAND® evaporating dish
PTFE, volume 100 mL
Synonym(s):
Evaporating dishes, PTFE
About This Item
UNSPSC Code:
41121812
Pricing and availability is not currently available.
material
PTFE
conical
feature
with spout
manufacturer/tradename
BRAND 45620
volume
100 mL
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)