Key Documents
BR386048
BRAND® Oxygen flask
volume 250-300 mL, with glass stopper NS 19/26
Synonym(s):
Bottles, Oxygen flasks, Winkler pattern
Select a Size
2 EA
$125.00
$125.00
Please contact Customer Service for Availability
Select a Size
Change View
2 EA
$125.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$125.00
Please contact Customer Service for Availability
material
soda-lime glass
packaging
pack of 2 ea
manufacturer/tradename
BRAND 386048
capacity
250-300 mL
volume
250-300 mL
General description
Soda-lime glass. For the determination of oxygen dissolved in water. The measured capacity is specified to ±0.01 mL. With labeling area. Solid, obliquely cut standard-ground glass stopper can be secured with a fastening clip. Each flask is calibrated with its own stopper, hence, stoppers and flasks are not interchangeable. Each flask and its stopper is marked with a unique matching identification number.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)