BR36552
BRAND® BLAUBRAND® volumetric flask PUR-coated
capacity 500 mL, joint: ST/NS 19/26
Synonym(s):
BLAUBRAND®, PURprotect, Volumetric flasks
Select a Size
$239.00
Select a Size
About This Item
$239.00
Recommended Products
material
glass
polypropylene stopper
Joint
joint: ST/NS19/26
feature
accuracy class A
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36552
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The PUR coating envelops the volumetric flasks like a protection skin. In the event of breakage, the safety coating helps contain the glass fragments. Compared to uncoated glass flasks the electrostatic charge is not increased. The light blue coating facilitates optical distinction. The maximum operating temperature at dry heat is 135°C (exposure time <30 min.).
Frequent autoclaving at 121°C reduces splintering protection. Cleaning up to max. 95°C.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service